Chlorite standard for ion chromatography, (CRM), Fluka™
Toimittaja: Honeywell Chemicals
E3010-100ML
HONCE3010-100MLEA 68.87 EUR
HONCE3010-100ML
Chlorite standard for ion chromatography, (CRM), Fluka™
Standardiaineet ja vertailumateriaalit Anionic Standards Chlorite Standards
Certified Reference Material (CRM)
  • Traceable to SRM from NIST
Sertifikaatit: ISO/IEC 17025:2017 (ANAB), ISO 17034:2016 (ANAB).