Concentrated Eluent for Ion Chromatography, Methanesulfonic Acid Solution
Toimittaja: Honeywell Chemicals
E2211-500ML
HONCE2211-500MLEA 170.77 EUR
HONCE2211-500ML
Concentrated Eluent for Ion Chromatography, Methanesulfonic Acid Solution
Standardiaineet ja vertailumateriaalit Anionic Standards Multi-Element Anioinic Standards
Certified Reference Material (CRM).
0.5 M Methanesulfonic acid in water