Multi-element solution 3 with Hg for ICP
Toimittaja: Honeywell Chemicals
E2178-100ML
HONCE2178-100MLEA 522.66 EUR
HONCE2178-100ML
Multi-element solution 3 with Hg for ICP
Standardiaineet ja vertailumateriaalit Cationic Standards Multi-Element Standards
Certified Reference Material (CRM).
10 mg/L each of Ag, Al, As, Ba, Be, Bi, Ca, Cd, Co, Cr, Cs, Cu, Fe, Ga, Hg, In, K, Li, Mg, Mn, Na, Ni, Pb, Rb, Se, Sr, Tl, U, V, Zn in HNO₃ 5%