Mixed calibration standard 3 for ICP
Toimittaja: Honeywell Chemicals
E2096-100ML
HONCE2096-100MLEA 337.38 EUR
HONCE2096-100ML
Mixed calibration standard 3 for ICP
Standardiaineet ja vertailumateriaalit Cationic Standards Multi-Element Standards
Certified Reference Material (CRM).
500mg/L As, 100mg/L Mo, 100mg/L Si in HNO₃ 2%