Rubidium ICP and ICP-MS Standards, (CRMs), Fluka™
Toimittaja: Honeywell Chemicals
E1343-100ML E1411-100ML
HONCE1343-100MLEA 96.66 EUR
HONCE1343-100ML HONCE1411-100ML
Rubidium ICP and ICP-MS Standards, (CRMs), Fluka™
Standardiaineet ja vertailumateriaalit Cationic Standards Rubidium Standards
Certified Reference Materials (CRMs)
  • Traceable to SRM from NIST
Sertifikaatit: ISO/IEC 17025:2017 (ANAB), ISO 17034:2016 (ANAB).