Iron ICP and ICP-MS Standards, (CRMs), Fluka™
Toimittaja: Honeywell Chemicals
E1318-100ML E1386-100ML
HONCE1318-100MLEA 110.48 EUR
HONCE1318-100ML HONCE1386-100ML
Iron ICP and ICP-MS Standards, (CRMs), Fluka™
Standardiaineet ja vertailumateriaalit Cationic Standards Iron Standards
Certified Reference Materials (CRMs)
  • Traceable to SRM from NIST
Sertifikaatit: ISO/IEC 17025:2017 (ANAB), ISO 17034:2016 (ANAB).