Barium ICP standards, (CRMs), Fluka™
Toimittaja: Honeywell Chemicals
E1008-100ML E1101-100ML E1135-100ML E1205-100ML
HONCE1008-100MLEA 77.57 EUR
HONCE1008-100ML HONCE1101-100ML HONCE1135-100ML HONCE1205-100ML
Barium ICP standards, (CRMs), Fluka™
Standardiaineet ja vertailumateriaalit Cationic Standards Barium Standards
Certified Reference Materials (CRMs)
  • Traceable to SRM from NIST
Sertifikaatit: ISO/IEC 17025:2017 (ANAB), ISO 17034:2016 (ANAB).