Manganese ICP standards, (CRMs), Fluka™
Toimittaja: Honeywell Chemicals
E1035-100ML E1111-100ML E1162-100ML
HONCE1035-100MLEA 77.57 EUR
HONCE1035-100ML HONCE1111-100ML HONCE1162-100ML
Manganese ICP standards, (CRMs), Fluka™
Standardiaineet ja vertailumateriaalit Cationic Standards Manganese Standards
Certified Reference Materials (CRMs)
  • Traceable to SRM from NIST
Sertifikaatit: ISO/IEC 17025:2017 (ANAB), ISO 17034:2016 (ANAB).