Tungsten ICP Standards, (CRMs), Fluka™
Toimittaja: Honeywell Chemicals
E1068-100ML E1127-100ML E1195-100ML E1262-100ML
HONCE1068-100MLEA 111.58 EUR
HONCE1068-100ML HONCE1127-100ML HONCE1195-100ML HONCE1262-100ML
Tungsten ICP Standards, (CRMs), Fluka™
Standardiaineet ja vertailumateriaalit Cationic Standards Tungsten Standards
Certified Reference Materials (CRMs)
  • Traceable to SRM from NIST
Sertifikaatit: ISO/IEC 17025:2017 (ANAB), ISO 17034:2016 (ANAB).