Antimony ICP Standards, (CRMs), Fluka™
Toimittaja: Honeywell Chemicals
E1119-100ML E1179-100ML E1247-100ML E1052-100ML
HONCE1119-100MLEA 95.97 EUR
HONCE1119-100ML HONCE1179-100ML HONCE1247-100ML HONCE1052-100ML
Antimony ICP Standards, (CRMs), Fluka™
Standardiaineet ja vertailumateriaalit Cationic Standards Antimony Standards
Certified Reference Materials (CRMs)
  • Traceable to SRM from NIST
Sertifikaatit: ISO/IEC 17025:2017 (ANAB), ISO 17034:2016 (ANAB).