Magnesium ICP standards, (CRMs), Fluka™
Toimittaja: Honeywell Chemicals
E1034-100ML E1161-100ML
HONCE1034-100MLEA 77.57 EUR
HONCE1034-100ML HONCE1161-100ML
Magnesium ICP standards, (CRMs), Fluka™
Standardiaineet ja vertailumateriaalit Cationic Standards Magnesium Standards
Certified Reference Materials (CRMs)
  • Traceable to SRM from NIST
Sertifikaatit: ISO/IEC 17025:2017 (ANAB), ISO 17034:2016 (ANAB).