Oxidation reagent 0.05 M I₂ in pyridine/water (90/10) (v/v), BioSyn™ for DNA/RNA synthesis, Burdick & Jackson™

Toimittaja: Honeywell Chemicals
Varoitus

BC664-1200KG BC664-2.5L BR664-4X2.5GL
HONCBC664-1200KGEA 60570 EUR
HONCBC664-1200KG HONCBC664-2.5L HONCBR664-4X2.5GL
Oxidation reagent 0.05 M I₂ in pyridine/water (90/10) (v/v), BioSyn™ for DNA/RNA synthesis, Burdick & Jackson™
Jodi

Burdick & Jackson™ BioSyn™ reagents & solvents for DNA/RNA synthesis


  • Honeywell Burdick & Jackson™ BioSyn™ Solvents and Reagents are specifically processed and purified to exacting specifications, ensuring outstanding synthesis efficiency
  • Ideal for oligonucleotide synthesis regardless of the application
  • All solvents and reagents are filtered (0.2-micron for oxidation reagents and 0.1-micron for all other reagents) to minimize particulate contamination

Kaava: I₂
Kiehumispiste: 115,25 °C (1.013 hPa.) (main component)
Sulamispiste: -42,2 °C (main component)
Tiheys: 0,983 g/cm3 (20 °C) (main component)
Leimahduspiste: 12,4 %(V) (main component)
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00011355
CAS-numero: 7553-56-2
UN: 1993
ADR: 3,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR