Capping Reagent, BioSyn™ for DNA/RNA synthesis, Burdick & Jackson™

Toimittaja: Honeywell Chemicals
Vaara

BC654-1100KG BC654-2.5L
HONCBC654-1100KGEA 0 EUR
HONCBC654-1100KG HONCBC654-2.5L
Capping Reagent, BioSyn™ for DNA/RNA synthesis, Burdick & Jackson™
Capping Reagent

Burdick & Jackson® BioSyn® reagents & solvents for DNA/RNA synthesis


  • Honeywell Burdick & Jackson® BioSyn® Solvents and Reagents are specifically processed and purified to exacting specifications, ensuring outstanding synthesis efficiency
  • Ideal for oligonucleotide synthesis regardless of the application
  • All solvents and reagents are filtered (0.2-micron for oxidation reagents and 0.1-micron for all other reagents) to minimize particulate contamination

Tiheys: 0,827 g/cm3 (20 °C)
UN: 2924
ADR: 3,II

Order Now

Testitulosten erittely

Appearance complying
Identity (IR) complying
Assay Of Acetonitrile 79.0 - 81.0 Vol.%
N-methylimidazole (GC) 19.0 - 21.0 Vol.%
Triazine Max. 20 ppm
Water (Karl Fischer) Max. 50 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR