Tubing for rotarus® peristaltic pumps, Tygon® 2001
Toimittaja: HIRSCHMANN
95214018 95214017 95214013 95214011 95214015 95214016 95214704 95214708 95214712 95214722 95214725
HIRS95214018EA 762 EUR
HIRS95214018 HIRS95214017 HIRS95214013 HIRS95214011 HIRS95214015 HIRS95214016 HIRS95214704 HIRS95214708 HIRS95214712 HIRS95214722 HIRS95214725
Tubing for rotarus® peristaltic pumps, Tygon® 2001
Letku Siirtoputki
These ultra-pure tube free of softeners for applications in the food industry and aggressive media with broad chemical resistance, free of softeners.
  • Temperature resistance from –78 to 57 °C
  • Hardness (Shore durometer A): 69