Multi-channel pipettes, mechanical, labopette®
Toimittaja: HIRSCHMANN
9475704 9475601 9475602 9475604 9475701 9475702
HIRS9475704EA 1220 EUR
HIRS9475704 HIRS9475601 HIRS9475602 HIRS9475604 HIRS9475701 HIRS9475702
Multi-channel pipettes, mechanical, labopette®
Pipetit ja pipetintäyttäjät Pipetit
The labopette® multi channel pipette for 96 microtiter plates features superior ergonomic characteristics, right down to the smallest detail.
  • Completely autoclavable without dismantling
  • Volume configuration with digital locking feature
  • Tip seal and safety filter to prevent contamination
  • Trigger grip ensures a firm but relaxed hold during pipetting
  • Tip ejection is deliberately separated from the pipetting button to avoid unintentional ejection
  • Special tension spring mechanism reduces the level of exertion required when compared to conventional pipettes