Dekantterit, matala malli, PP
Toimittaja: HIRSCHMANN
7600186 7600160 7600170 7600175 7600180 7600183 7600189 7600190 7600191 7600193 7600194 7600196 7600197
HIRS7600186EA 26.8 EUR
HIRS7600186 HIRS7600160 HIRS7600170 HIRS7600175 HIRS7600180 HIRS7600183 HIRS7600189 HIRS7600190 HIRS7600191 HIRS7600193 HIRS7600194 HIRS7600196 HIRS7600197
Dekantterit, matala malli, PP
Dekantterit
PP, highly translucent.
  • Very good chemical resistance
  • Cleaning up to maximum +60 °C is recommended to preserve marks and inscriptions
  • For autoclaving (+121 °C) beakers with an embossed scale should be selected
With printed blue scale, beaded rim and spout.
Sertifikaatit: ISO 7056