Mittalasit, PP, luokka B
Toimittaja: HIRSCHMANN
7500160 7500170 7500175 7500180 7500186 7500190 7500193 7500194 7501160 7501170 7501175 7501180 7501186 7501190 7501193 7501194
HIRS7500160EA 52.1 EUR
HIRS7500160 HIRS7500170 HIRS7500175 HIRS7500180 HIRS7500186 HIRS7500190 HIRS7500193 HIRS7500194 HIRS7501160 HIRS7501170 HIRS7501175 HIRS7501180 HIRS7501186 HIRS7501190 HIRS7501193 HIRS7501194
Mittalasit, PP, luokka B
Mittalasit Measuring Cylinders
PP, translucent, tall form, class B.
  • Calibrated to 'In'
  • Hexagonal base, ring mark on the principal points
  • Thermal stresses of up to +80 °C do not result in the tolerance limit being permanently exceeded
Sertifikaatit: DIN 12681 B / ISO 6706