Mittapullo, luokka A
Toimittaja: HIRSCHMANN
2822166 2822153 2822161 2822165 2822170 2822175 2822178 2822180 2822181 2822185 2822186 2822190 2822193 2822194 2822152 2822160 2822171 2822192 2822197 2822198
HIRS2822166EA 24.6 EUR
HIRS2822166 612-1575 612-1576 612-1577 612-1578 612-1579 612-1580 612-1581 612-1582 612-1583 612-1584 612-1585 612-1586 612-1587 HIRS2822152 HIRS2822160 HIRS2822171 612-5739 612-5740 612-5742
Mittapullo, luokka A
Kolvit ja erlenmeyerit Mittapullot
DURAN®, borosilicate glass, with PE stopper, conformity certified (DE-M marking).
  • Amber stain markings
  • Certificate of conformity with dated batch identification available via website: (www.hirschmannlab.de)
*Wide neck
Sertifikaatit: DIN EN ISO 1042