Avantor® Hichrom, HPLC Columns, RPB
Toimittaja: Avantor
HI-10RPB-100PEA 0 EUR
HI-10RPB-100P HICHHI-10RPB-10CP HI-10RPB-150P HI-10RPB-250P HI-10RPB-50P HI-10RPB-100SP1 HI-10RPB-150SP1 HI-10RPB-250SP1 HIRPB-100A HIRPB-100AS HIRPB-100P HICHHIRPB-10CP HIRPB-150A HIRPB-150AS HIRPB-150P HIRPB-250A HIRPB-250AM HIRPB-250AS HIRPB-250P HIRPB-250SP HIRPB-50P HI-10RPB-100P3 HI-10RPB-10CP3 HI-10RPB-150P3 HI-10RPB-250P3 HI-10RPB-50P3 HI-10RPB-50SP1 HI-3.5RPB100AF HI-3.5RPB100AS HI-3.5RPB150AM HI-3.5RPB150AS HIRPB-100P3 HIRPB-10C5 HIRPB-150P3 HIRPB-250P3 HIRPB-50P3 HI3.5RPB10CM5 HIRPB-10C5-SK
Avantor® Hichrom, HPLC Columns, RPB
Kromatografiakolonnit
Avantor® Hichrom RPB is a high purity base deactivated silica which offers a unique selectivity due to the proprietary C8/C18 multi-alkyl bonding and exhaustive endcapping employed. The range includes 3,5, 5 and 10 μm phases, which are suitable for LC-MS applications and preparative separations respectively.
  • Increased productivity due to reduced analysis time
  • Decreased column length without sacrificing efficiency
  • Excellent chromatography
Avantor® Hichrom RPB is specifically designed for the reversed-phase separation of basic molecules. Residual acidicMore Product Information