Avantor® ACE® HILIC-N, HPLC/UHPLC Columns, Analytical
Toimittaja: Avantor
HILN-17-0502UEA 1100 EUR
HILN-17-0502U HILN-17-0503U HILN-17-7502U HILN-17-1002U HILN-17-1003U HILN-3-0102GD HILN-3-0303U HILN-3-0502U HILN-3-0503U HILN-3-0546U HILN-3-7503U HILN-3-1002U HILN-3-1003U HILN-3-1046U HILN-3-1502U HILN-3-1503U HILN-3-1546U HILN-3-2502U HILN-3-2503U HILN-3-2546U HILN-5-0103GD HILN-5-0502U HILN-5-0503U HILN-5-0546U HILN-5-1002U HILN-5-1003U HILN-5-1046U HILN-5-1502U HILN-5-1503U HILN-5-1546U HILN-5-2502U HILN-5-2503U HILN-5-2546U HILN-5-7530 HILN-5-7520 HILN-5-3030 HILN-5-3020 HILN-5-2530 HILN-5-2520 HILN-5-1530 HILN-5-1520 HILN-5-1510 HILN-5-1220 HILN-5-1030 HILN-5-1020 HILN-5-1010 HILN-5-0530 HILN-5-0520 HILN-5-0510 ACETHILN-3-0103GD HILN-17-GD2U HILN-17-D1002
Avantor® ACE® HILIC-N, HPLC/UHPLC Columns, Analytical
Kromatografiakolonnit
Avantor® ACE® HILIC-N columns have been designed specifically for HILIC applications and are especially useful for the separation of polar analytes. These columns are part of a suit of 3 proprietary HILIC columns (HILIC-A, HILIC-B and HILIC-N) developed to enable chromatographers to achieve reproducible and successful HILIC separations using a systematic approach to method development.
  • Proprietary polyhydroxy neutral character HILIC phase
  • Compatible with all manufacturers’ HPLC/UHPLC systems
  • Excellent column performance, inertness and robustness
  • Simplified