Avantor® ACE® HILIC-B, HPLC/UHPLC Columns, Analytical
Toimittaja: Avantor
HILB-3-0303UEA 857 EUR
HILB-3-0303U HILB-3-0502U HILB-3-0503U HILB-3-0546U HILB-3-1002U HILB-3-1003U HILB-3-1046U HILB-3-1502U HILB-3-1503U HILB-3-1546U HILB-3-2502U HILB-3-2503U HILB-3-2546U HILB-5-0103GD HILB-5-0502U HILB-5-0503U HILB-5-0546U HILB-5-1002U HILB-5-1003U HILB-5-1046U HILB-5-1502U HILB-5-1503U HILB-5-1546U HILB-5-2502U HILB-5-2503U HILB-5-2546U HILB-17-0203U HILB-17-0302U HILB-17-0303U HILB-17-0502U HILB-17-0503U HILB-17-7502U HILB-17-1002U HILB-17-1003U HILB-3-0102GD HILB-3-0103GD HILB-3-0302U HILB-17-GD2U HILB-17-D1002
Avantor® ACE® HILIC-B, HPLC/UHPLC Columns, Analytical
Kromatografiakolonnit
Avantor® ACE® HILIC-B columns have been designed specifically for HILIC applications and are especially useful for the separation of polar analytes. These columns are part of a suit of 3 proprietary HILIC columns (HILIC-A, HILIC-B and HILIC-N) developed to enable chromatographers to achieve reproducible and successful HILIC separations using a systematic approach to method development.
  • Proprietary aminopropyl basic character HILIC phase
  • Compatible with all manufacturers’ HPLC/UHPLC systems
  • Excellent column performance, inertness and robustness
  • Simplified