Avantor® Hichrom, Esikolonnit ja fritit, esikolonnin suodattimet
Toimittaja: Avantor
554-7720EA 78.4 EUR
554-7720 554-7719 HI-685 554-7722 HI-615 HI-686 HICHHI-602 HICHHI-602X
Avantor® Hichrom, Esikolonnit ja fritit, esikolonnin suodattimet
Suodattimet Esikolonnit ja fritit
Placed immediately before the column, the precolumn filter traps particles introduced with the sample. For samples, which may irreversibly absorb onto the column, the use of guard cartridges is strongly recommended.
  • Protect columns from particulates, ultra low dead volume
  • Virtually no dead volume with no band broadening or loss of efficiency
  • Compatible with all HPLC columns
Filter HI-704 comprises a stainless steel holder with a 0,020" thru hole and a 2 µm PEEK encapsulated stainless steel frit (supplied).
The fingertight HI-685 precolumn filterMore Product Information