Avantor® Hichrom PEEKsil™, Chromatography Tubing, Stainless Steel
Toimittaja: Avantor
HICHHI-1109EA 126 EUR
HICHHI-1109 HICHHI-1128
Avantor® Hichrom PEEKsil™, Chromatography Tubing, Stainless Steel
Letku Chromatography Tubing
Tubings are made of 316 stainless steel.
  • Resistant to chemicals
The precision bore is 1/16" type tubing.