Avantor® Alltima HP, HPLC columns, HILIC, 5 µm
Toimittaja: Avantor
HICH86465EA 1100 EUR
HICH86465 HICH86464 HICH86466 86479/N 86480/N
Avantor® Alltima HP, HPLC columns, HILIC, 5 µm
Kromatografiakolonnit
Avantor® Alltima HP columns features hydrophilic interaction chromatography that uses small amounts of water for increased sensitivity with microbore applications.
  • High resolution for complex samples
  • High purity silica
  • Excellent column stability
These columns combine the selectivity and performance needed to overcome the most challenging separation needs. The low column bleed makes these columns ideal for microbore applications.
These are widely used for polar analytes that are difficult to retain by reversed-phase.