Avantor® Alltima, HPLC Columns, C8
Toimittaja: Avantor
HICH81392EA 0 EUR
HICH81392 HICH81397 HICH81398 HICH88372 HICH88072 HICH88073 HICH88076 HICH88077 96081/N
Avantor® Alltima, HPLC Columns, C8
Kromatografiakolonnit
Avantor® Alltima HPLC columns are high quality phases designed for general purpose applications.
  • Base deactivated silica phases
  • Stable bonding for long column lifetime
  • Multiple selectivity options
  • USP L7
Lower capacity compared to C18 phases. Recommended for reversed phase applications where C18 is too retentive.