Avantor® Ultrasphere®, HPLC-kolonni
Toimittaja: Avantor
HICH235330EA 1440 EUR
HICH235330 HICH235329 HICH235332 HICH235333 HICH235334 HICH235335 HICH235341 HICH235342 HICH237390 HICH237520 HICH237521 HICH237573 HICH237574 HICH240002 HICH244046 HICH244070 HICH244071 HICH244254 HICH244434 243533N 243534N 235327.
Avantor® Ultrasphere®, HPLC-kolonni
Kromatografiakolonnit
Avantor® Ultrasphere® columns remain widely referenced within both industry and academia and are recognised to provide excellent chromatographic performance.
  • 3 and 5 μm spherical porous silica
  • High reproducibility, narrow particle size distribution
  • Optimal surface coverage for long column lifetime
Avantor® Ultrasphere® ODS (3 µm) and Octyl phases are available in both standard analytical column hardware and a high efficiency Ultrasphere-XL High Speed cartridge column design, which includes an integral cartridge column holder assembly.