Avantor® Hichrom HI-DEX G-01, GC Columns
Toimittaja: Avantor
HI74-25-015-15EA 1250 EUR
HI74-25-015-15 HI74-25-015-25 HI74-25-015-30 HI74-25-015-50 HI74-25-015-60 HI74-25-025-15 HI74-25-025-25 HI74-25-025-30 HI74-25-025-50 HI74-25-025-60 HI84-10-010-10 HI84-10-010-5 HIGCHI74-15-015-30
Avantor® Hichrom HI-DEX G-01, GC Columns
Kromatografiakolonnit
Unique selectivity for the separation of enantiomers. Useful for the analysis of chiral compounds in fragrances, pesticides and pharmaceuticals in which the determination of the ratio of enantiomers in a sample is necessary.
Max temperature limits may change depending on the film thickness.