Avantor® Hichrom, Column Frits
Toimittaja: Avantor
HI-120EA 20.4 EUR
HI-120 HI-145 HICHHI-100 HICHHI-104 HICHHI-105 HICHHI-126 HICHHI-133
Avantor® Hichrom, Column Frits
Suodattimet Esikolonnit ja fritit
Frit made of 316 grade stainless steel.
  • Helps in controlling porosity
A 2 μm porosity frit is recommended for retention of 5 to 20 μm packing material and 0,5 μm porosity frit is recommended for 3 to 4 μm packing materials.
Stainless steel with encapsulated PEEK used to prevent sample and eluent from entering dead volume space between the end fitting and also with the top of the column. This results in improved peak symmetry with 1,0 to 3,2 mm Int.Ø columns.