Capillaries for LC systems
Toimittaja: AGILENT HEWLETT PACKARD
01090-87610 5021-1823 G1315-87311 G1316-87300 5067-4748 5067-4658 5067-5107 5067-5108 5067-4669 5067-4735 5067-4744 5067-4746 5067-5104
HEWL01090-87610EA 0 EUR
HEWL01090-87610 HEWL5021-1823 HEWLG1315-87311 HEWLG1316-87300 HEWL5067-4748 HEWL5067-4658 HEWL5067-5107 HEWL5067-5108 HEWL5067-4669 HEWL5067-4735 HEWL5067-4744 HEWL5067-4746 HEWL5067-5104
Capillaries for LC systems
Letku Chromatography Tubing
Stainless steel capillaries are made of flexible stainless steel (0,6 mm OD) with ¹/₁₆" OD tubing at both ends to accept standard fittings.
Pre-swaged fittings are assembled according to Swagelok specifications.