Ultra low temperature freezers, with 5 inner doors –40 °C, TDE series
Toimittaja: Thermo Scientific
TDE40040FV5ID TDE50040FV5ID TDE60040FV5ID
HERATDE40040FV5IDEA 12750 EUR
HERATDE40040FV5ID HERATDE50040FV5ID HERATDE60040FV5ID
Ultra low temperature freezers, with 5 inner doors –40 °C, TDE series
Pakastimet Ultra Low Temperature Freezers
Thermo Scientific™ TDE Series ‒40 °C ultra-low temperature freezers, powered by H-drive, feature three upright models with 5 inner doors, maximizing storage capacity from 40000 up to 60000 2 ml vials. The TDE Series is a sustainable and reliable solution with ultimate sample security and operational savings for every laboratory.
  • Powered by H-drive, save up to 25% in energy usage and heat rejection over conventional refrigerant ultra-low freezers
  • Easy-to-use set-up wizard, event log, temperature mapping and data downloads
  • Capacitive touch even when using