Accessories for freezers, CryoMed™ and CryoPlus™
Toimittaja: Thermo Fisher Scientific
4000566 4000320 4000385 4000701 4000702 4000705 4000706 4000386 4000303 4000700 4000707 185089 4000704 4000708 4000703 4000294 4000314 4000555
THBR4000566EA 224 EUR
THBR4000566 HERA4000320 HERA4000385 HERA4000701 HERA4000702 HERA4000705 HERA4000706 HERA4000386 HERA4000303 HERA4000700 HERA4000707 HERA185089 HERA4000704 HERA4000708 HERA4000703 THBR4000294 THBR4000314 THBR4000555
Accessories for freezers, CryoMed™ and CryoPlus™
Pakastimet Cryogenic Freezers
More flexibility for your controlled-rate freezing applications with Thermo Scientific™ racks for the CryoMed™ controlled-rate freezer. Includes an assortment of cane, canister, straw and vial racks.
  • Thermocouple sensors for cryo tubes, sheath straw and vial samples, includes male plug
  • Warranty varies by country, please contact us for details
Perform controlled-rate freezing applications with a controlled-rate freezer and these Thermo Scientific™ CryoMed™ bag freezing presses.
Improve the functionality of Thermo Scientific™ Cryomed™ controlled-rateMore Product Information
Order Now
TEKNISET TIEDOT
Tuotenro THBR4000566