Racks for Revco™ ExF, DxF and HERAfeeze™ HFU B Freezers
Toimittaja: THERMO SAVANT
398326 1950728
THSA398326EA 279 EUR
THSA398326 HERA1950728
Racks for Revco™ ExF, DxF and HERAfeeze™ HFU B Freezers
Koeputkitelineet Telineet kryolaatikoille
Maximize the storage space in your ultra-low temperature freezer with these racks.
Includes sliding drawer racks, adjustable side access box racks and microplate racks in standard or deepwell formats.