Demountable cuvettes
Toimittaja: HELLMA
106-0.50-40 106-0.10-40 106-0.20-40 106-0.01-40 124-0.1-40 124-0.01-40 124-0.2-40 124-0.5-40
634-9235EA 381 EUR
634-9235 634-9237 634-9239 HELL106-0.01-40 HELL124-0.1-40 HELL124-0.01-40 HELL124-0.2-40 HELL124-0.5-40
Demountable cuvettes
Kyvetit Spektrofotometrin kyvetit
Demountable cuvettes for short pathlength being easier to clean used in microscopic examinations.
Suitable for high precision analytical techniques.
Tilaustiedot: A cell holder is usually required for use with photometers.