Sealable Quartz Glass Cells

Toimittaja: HELLMA
117100F-10-40 117104-10-40 117100-10-40 117104F-10-40 117200-10-40 117200F-10-40 117104F-10K-40
HELL117100F-10-40EA 616 EUR
HELL117100F-10-40 634-9541 634-9542 HELL117104F-10-40 HELL117200-10-40 HELL117200F-10-40 HELL117104F-10K-40
Sealable Quartz Glass Cells
Kyvetit Spektrofotometrin kyvetit

Hellma® high precision quartz glass macro cells for absorption measurements in the UV/Vis range, 10 mm path length, with open screw cap.



  • Cells made of high performance quartz glass

  • Wavelength range from 200 to 2500 nm
  • With open screw cap
  • Silicone seal
  • Volume: 1400 to 3500 μl 


Hellma® precision cells are 100% manufactured in Germany and are suitable for all common spectrophotometers.



Highest precision: Hellma® precision cells are characterized by the highest precision in path length accuracy and parallelism. This guarantees continuously

More Product Information
Order Now

TEKNISET TIEDOT

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR