PharmaFocus® Premium Peristaltic Pump Tubing
Toimittaja: Freudenberg Medical Europe GmbH
49464761 49464762 49446460 49464765 49446463 49464800 49456985 49464764 49457003 49464766 49463403 49443818 49464802 49464804 49464767 49464801 49464803 49464768 494555845
HELI49464761EA 183 EUR
HELI49464761 HELI49464762 HELI49446460 HELI49464765 HELI49446463 HELI49464800 HELI49456985 HELI49464764 HELI49457003 HELI49464766 HELI49463403 HELI49443818 HELI49464802 HELI49464804 HELI49464767 HELI49464801 HELI49464803 HELI49464768 HELI494555845
PharmaFocus® Premium Peristaltic Pump Tubing
Letku Siirtoputki
PharmaFocus® Premium Peristaltic Pump Tubing is a high purity silicone tubing specifically designed for peristaltic pumps and pinch valves and is ideal for demanding pumping applications. The Peristaltic Pump Tubing undergoes a stabilization process unique to Freudenberg Medical that provides the additional benefits of superior strength, performance, and longer life.
  • Performs in high purity fluid transfer and pump applications
  • Tube resilience and adaptability due to excellent flexibility
  • High visibility of fluid transport
  • Versatile processing with cut-to-