Electronic max/min thermometer
Toimittaja: GLASWARENFABRIK KARL HECHT
43656010
HECH43656010EA 26.9 EUR
HECH43656010
Electronic max/min thermometer
Lämpömittarit Digital Thermometers
Maximum-minimum electronic thermometer used for measuring indoor and outdoor temperature.
  • Present temperature/highest and lowest day temperature
  • For hanging or standing
  • Extra thin cable for easily leading through window seals
  • Waterproof sensor for outdoor measurements
* Indoor
** Outdoor
Toimitustiedot: Incl. 1,5 V AA battery.