Tanks for water baths
Toimittaja: GLASWARENFABRIK KARL HECHT
43187003 43194001
HECH43187003EA 183 EUR
HECH43187003 HECH43194001
Tanks for water baths
Nestesäiliöt ja astiat Nelikulmaiset säiliöt
Tubs made of Plexiglas®, available with or without sloping base. Sloping base facilitates circulation of the liquid.
For use with water bath thermostats.