Test tubes, flat bottom, without rim, heavy wall
Toimittaja: GLASWARENFABRIK KARL HECHT
42779070 42779065 92277901450
HECH42779070EA 35.8 EUR
HECH42779070 HECH42779065 HECH92277901450
Test tubes, flat bottom, without rim, heavy wall
Putket Koeputket
Made of AR-Glas® (soda glass).
  • Heavy wall
  • Flat bottom
  • Good chemical and physical properties