Reduction and expansion adapters for conical ground joints
Toimittaja: GLASWARENFABRIK KARL HECHT
42387001 42387002 42387003 42387004 42387005 42387006 42387008 42387009 42387010 42387011 42387012 42387014
HECH42387001EA 17.3 EUR
HECH42387001 HECH42387002 HECH42387003 HECH42387004 HECH42387005 HECH42387006 HECH42387008 HECH42387009 HECH42387010 HECH42387011 HECH42387012 HECH42387014
Reduction and expansion adapters for conical ground joints
Sovittimet Sovittimet, NS-hios
Borosilicate glass 3,3
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
Reduction adapter from large cone to small socket or expansion adapter from small cone to large socket.