Dropping bottles, glass
Toimittaja: GLASWARENFABRIK KARL HECHT
41314060 41314010 41314020 41314050
HECH41314060EA 75.7 EUR
HECH41314060 HECH41314010 HECH41314020 HECH41314050
Dropping bottles, glass
Pullot Tippapullo
Made of amber glass.
  • Good chemical and physical properties
  • For use in applications that do not involve high thermal stresses
  • Amber coloured to protect light sensitive substances
Dropping bottles without cap, with thread DIN 18.
Tilaustiedot: Pipette kit and screw cap must be ordered seperately.