Vacuum desiccators, Novus
Toimittaja: GLASWARENFABRIK KARL HECHT
41160010 41160015 41160020 41160025 41160030
HECH41160010EA 159 EUR
HECH41160010 HECH41160015 HECH41160020 HECH41160025 HECH41160030
Vacuum desiccators, Novus
Eksikaattorit ja kaappieksikaattorit Eksikaattori, lasia
Vacuum desiccators are made of borosilicate glass 3.3 and are tested for tension, vacuum tightness and pressure.
Sertifikaatit: DIN 12491.