Graduated pipettes, conformity certified, partial delivery
Toimittaja: GLASWARENFABRIK KARL HECHT
40545730 40545736
HECH40545730EA 150 EUR
HECH40545730 HECH40545736
Graduated pipettes, conformity certified, partial delivery
Pipetit ja pipetintäyttäjät Pipetit, asteikolla
Glass.
  • Enzyme test pipettes with long, tapered, sturdy tip
  • Precision type, for partial delivery (not calibrated to tip), calibrated to deliver (Ex),
  • Zero at top, marked main points
  • Tolerance indicated on each pipette, amber graduations