Bottles and tube racks
Toimittaja: HEATHROW
HS120085 HS120086
HEATHS120085EA 36.7 EUR
HEATHS120085 HEATHS120086
Bottles and tube racks
Koeputkitelineet Pullotelineet
Wire, epoxy coated.
  • Rack suitable for Bijou bottles and low profile tubes up to 25 mm diameter
  • Mesh bottom support
  • Chemically-resistant steel wire racks
For transporting and handling samples.