Pipette holder, Flip&Grip®
Toimittaja: HEATHROW
HS120026 HS120056
HEATHS120026EA 181 EUR
HEATHS120026 HEATHS120056
Pipette holder, Flip&Grip®
Pipettitelineet
The unique pipette holder gives you the option of using the pipette holder on the bench top or mount the holder under a shelf or counter to help maximize your working space.
  • Holds most common brands
  • Rubber grips prevent scratching pipette
  • Holder rotates for easy pipette selection and storage
  • Weighted base
  • Pipettes can be held in the rubber grips and/or by the plunger hooks
  • Hardware provided to install under shelf
Will hold up to 12 single-channel pipettes, or a combo of 3 single- and 3 multichannel pipettes.
W×H under shelf style: 193×187 mm
W×HMore Product Information