Test sieves
Toimittaja: NEXOPART
206136800 206159564 206136985 206158604 206158598 206158390 206158574 206158567 206158543 206158536 206158512 206158505 206158482 206158451 206158406 206158666 206158376 206158369 206158345 206158338 206158307 206158284 206158277 206158253 206158246 206158222 206158215 206158420 206158895 206159144 206159120 206159113 206159090 206159083 206159069 206159052 206159014 206159007 206158987 206158970 206158956 206158949 206158635 206158772 206159151 206158680 206158710 206158727 206158925 206158758 206158918 206158789 206158802 206158833 206158857 206158871 206158659 206158741 206137449 206137807 206136022 206136077 206136237 206136572 206136633 206136770 206136787 206137159 206137371 206137609 206137616 206158819 206159281 206135612 206158697 206158314 206158437 206158628 206136008 206134943 206135520 206134752 206134837 206134875 206134899 206134912 206134936 206134950 206134974 206134981 206135001 206135018 206135032 206135049 206135063 206135087 206135094 206135117 206135124 206135148 206135155 206135179 206135186 206135209 206135216 206135230 206135247 206135261 206135278 206135292 206135308 206135315 206135339 206135346 206135360 206135377 206135391 206135414 206135438 206135445 206135469 206135476 206135490 206135506 206135544 206135551 206135575 206135582 206135605 206135629 206135643 206135650 206135674 206135681 206135704 206135711 206135735 206135742 206135766 206135803 206135889 206135896 206135902 206135926 206135940 206135964 206136343 206136466 206159311 206159588 206159922
HAVE206136800EA 167 EUR
HAVE206136800 HAVE206159564 HAVE206136985 HAVE206158604 HAVE206158598 HAVE206158390 HAVE206158574 HAVE206158567 HAVE206158543 HAVE206158536 HAVE206158512 HAVE206158505 HAVE206158482 HAVE206158451 HAVE206158406 HAVE206158666 HAVE206158376 HAVE206158369 HAVE206158345 HAVE206158338 HAVE206158307 HAVE206158284 HAVE206158277 HAVE206158253 HAVE206158246 HAVE206158222 HAVE206158215 HAVE206158420 HAVE206158895 HAVE206159144 HAVE206159120 HAVE206159113 HAVE206159090 HAVE206159083 HAVE206159069 HAVE206159052 HAVE206159014 HAVE206159007 HAVE206158987 HAVE206158970 HAVE206158956 HAVE206158949 HAVE206158635 HAVE206158772 HAVE206159151 HAVE206158680 HAVE206158710 HAVE206158727 HAVE206158925 HAVE206158758 HAVE206158918 HAVE206158789 HAVE206158802 HAVE206158833 HAVE206158857 HAVE206158871 HAVE206158659 HAVE206158741 HAVE206137449 HAVE206137807 HAVE206136022 HAVE206136077 HAVE206136237 HAVE206136572 HAVE206136633 HAVE206136770 HAVE206136787 HAVE206137159 HAVE206137371 HAVE206137609 HAVE206137616 HAVE206158819 HAVE206159281 HAVE206135612 HAVE206158697 HAVE206158314 HAVE206158437 HAVE206158628 HAVE206136008 HAVE206134943 HAVE206135520 HAVE206134752 HAVE206134837 HAVE206134875 HAVE206134899 HAVE206134912 HAVE206134936 HAVE206134950 HAVE206134974 HAVE206134981 HAVE206135001 HAVE206135018 HAVE206135032 HAVE206135049 HAVE206135063 HAVE206135087 HAVE206135094 HAVE206135117 HAVE206135124 HAVE206135148 HAVE206135155 HAVE206135179 HAVE206135186 HAVE206135209 HAVE206135216 HAVE206135230 HAVE206135247 HAVE206135261 HAVE206135278 HAVE206135292 HAVE206135308 HAVE206135315 HAVE206135339 HAVE206135346 HAVE206135360 HAVE206135377 HAVE206135391 HAVE206135414 HAVE206135438 HAVE206135445 HAVE206135469 HAVE206135476 HAVE206135490 HAVE206135506 HAVE206135544 HAVE206135551 HAVE206135575 HAVE206135582 HAVE206135605 HAVE206135629 HAVE206135643 HAVE206135650 HAVE206135674 HAVE206135681 HAVE206135704 HAVE206135711 HAVE206135735 HAVE206135742 HAVE206135766 HAVE206135803 HAVE206135889 HAVE206135896 HAVE206135902 HAVE206135926 HAVE206135940 HAVE206135964 HAVE206136343 HAVE206136466 HAVE206159311 HAVE206159588 HAVE206159922
Test sieves
Seulat Testisiivilät
These ALPINE air jet test sieves are suitable for fully automatic sieve recognition. These test sieves are made of brass/stainless steel frame with metal wire cloth made from stainless steel.
Sertifikaatit: According to EN ISO/IEC 17025, DIN EN 10204.
Toimitustiedot: Supplied with serial number, inspection certificate and calibration certificate on request.