Kromatografian ruiskut, Digital Syringes™
Toimittaja: HAMILTON BONADUZ
DS80230 DS80930 DS80300 DS80400 DS80365 DS81030 DS80030 DS80530 DS80075 DS80100 DS80135 DS80200 DS80275 DS80330 DS80383 DS80430 DS80800 DS80500 DS80000 DS80565 DS80600 DS80608 DS80630 DS80665 DS80700 DS80730 DS80465 DS80830 DS88011 DS88400 DS88411 DS88500 DS88511 DS80765 DS87900 DS81176 DS80865 DS80900 DS80975 DS81000 DS81075 DS81100 DS88000 DS81175 DS87930 DS81217 DS81230 DS86200 DS86211 DS86250 DS86259 DS81130
HAMIDS80230EA 2350 EUR
HAMIDS80230 HAMIDS80930 549-1280 549-1283 549-1282 HAMIDS81030 HAMIDS80030 549-1287 HAMIDS80075 HAMIDS80100 HAMIDS80135 HAMIDS80200 HAMIDS80275 549-1281 HAMIDS80383 549-1284 549-1295 549-1286 HAMIDS80000 549-1288 549-1289 HAMIDS80608 549-1290 549-1291 549-1292 549-1293 549-1285 549-1296 HAMIDS88011 HAMIDS88400 HAMIDS88411 HAMIDS88500 HAMIDS88511 549-1294 549-1298 HAMIDS81176 549-1297 HAMIDS80900 HAMIDS80975 HAMIDS81000 HAMIDS81075 HAMIDS81100 HAMIDS88000 HAMIDS81175 549-1299 HAMIDS81217 HAMIDS81230 HAMIDS86200 HAMIDS86211 HAMIDS86250 HAMIDS86259 HAMIDS81130
Kromatografian ruiskut, Digital Syringes™
Ruiskut Ruiskut kromatografiaan
The Digital Syringe™ is a calibrated device composed of a serialised syringe and a serialised digital electronic holder.
  • Achieve precision up to 0,5%
  • Dispense volumes from 500 µl down to 50 nl
  • Device also reduces the possibility of plunger bending
A digital readout allows enhanced syringe volume readability, while an adjustable plunger stop assures accurate, reproducible sample injections.
Toimitustiedot: Each unit is delivered with a certificate of calibration.