GC septa, low bleed high temperature
Toimittaja: HAMILTON BONADUZ
75839 75837 75838 75840 75841 75834 75836 75835 75833
HAMI75839EA 198 EUR
HAMI75839 HAMI75837 HAMI75838 HAMI75840 HAMI75841 HAMI75834 HAMI75836 HAMI75835 HAMI75833
GC septa, low bleed high temperature
Septumi
Low bleed high temperature septa are designed for the lowest bleed possible at temperatures of up to 400 °C. They can be used in combination with Mass Spectrometers or in applications where septa bleed interferes with trace analysis. These septa are typically used for manual injection applications.
  • Extended temperature range, low-bleed
  • Virtually eliminates injection-port sticking
  • Each batch GC-FID tested
  • Ideal for use with low-bleed, 'Mass Spec' capillary columns
The 5, 9, 11 and 11,5 mm diameter septa are available with a center depression whichMore Product Information