Single channel pipettes, mechanical, fixed / variable volume, SoftGrip™
Toimittaja: HAMILTON BONADUZ
55019-36 55019-32 55019-38 55019-40 55019-11 55019-13 55019-15 55019-17 55019-19 55019-30 55019-34 55019-01 55019-03 55019-05 55019-07 55019-09
HAMI55019-36EA 401 EUR
HAMI55019-36 HAMI55019-32 HAMI55019-38 HAMI55019-40 HAMI55019-11 HAMI55019-13 HAMI55019-15 HAMI55019-17 HAMI55019-19 HAMI55019-30 HAMI55019-34 HAMI55019-01 HAMI55019-03 HAMI55019-05 HAMI55019-07 HAMI55019-09
Single channel pipettes, mechanical, fixed / variable volume, SoftGrip™
Pipetit ja pipetintäyttäjät Pipetit
Thanks to a soft contoured handle and a curved hilt these pipettes rest comfortable in your hand. The slim ejection sleeve is made from a high strength, chemically resistant polymer. The slim design allows the pipette to reach to the bottom of narrow tubes without the need for extended length tips.
  • Built from high quality, state-of-the-art materials designed to provide years of reliable service
  • All pipettes are completely autoclavable for easy decontamination and sterilisation without the need for disassembly and recalibration
  • Isolated volume ring prevents