Lisävarusteet, neulan puhdistusvälineet
Toimittaja: HAMILTON BONADUZ
18311 76620 18310 18300 18301 18302 18303 18304 18306
549-1250EA 120 EUR
549-1250 549-1251 549-1249 549-1244 549-1245 549-1246 549-1247 549-1248 HAMI18306
Lisävarusteet, neulan puhdistusvälineet
Puhdistusaineet Pesuaineet ja pintojen puhdistusaineet
To clean syringes it is best to use a solvent known to be effective in solvating. Ideally the solvent would be non-alkaline, non-phosphate, and non-detergent based.
  • Biodegradable, non-phosphate, cleaning concentrate that is suitable for many common sample types
  • For some needle obstructions, mechanical cleaning may be necessary
For this purpose various sized tungsten cleaning wires can be used. They can be threaded through the needle to clear a blockage.
To clean syringes it is best to use a solvent known to be effective in solvating. Ideally theMore Product Information