Refrigerated Circulators, ARCTIC
Toimittaja: THERMO ELECTRON LED
152-5281 153-5281 153-5401 153-5101 153-4281 153-4101 152-4281 153-4051 156-5281 156-5101 156-4281 156-4101 156-4051 152-4051 155-4101 155-5101 152-4101 152-5101 1555101 1554101 1555251 1565101 1564101 1525101 1524101 1525251 1524251 1535101 1534101 1535251 1534251
HAKA152-5281EA 0 EUR
HAKA152-5281 HAKA153-5281 HAKA153-5401 HAKA153-5101 HAKA153-4281 HAKA153-4101 HAKA152-4281 HAKA153-4051 HAKA156-5281 HAKA156-5101 HAKA156-4281 HAKA156-4101 HAKA156-4051 HAKA152-4051 HAKA155-4101 HAKA155-5101 HAKA152-4101 HAKA152-5101 HAKA1555101 HAKA1554101 HAKA1555251 HAKA1565101 HAKA1564101 HAKA1525101 HAKA1524101 HAKA1525251 HAKA1524251 HAKA1535101 HAKA1534101 HAKA1535251 HAKA1534251
Refrigerated Circulators, ARCTIC
Kiertotermostaatit Suljetut termostaattihaudejärjestelmät Jäähdyttävät termostaattihauteet
CFC-free refrigerated bath circulators provide cooling for open and closed applications. These bath circulators with a stainless-steel reservoir can be configured with six different controller options, so the user can choose the right model for the application.
  • Energy-saving feature limits power consumption
  • Three-sided ventilation: Up to two sides can be blocked with full refrigeration performance with no degradation of equipment
  • Features draining port at the front and integrated handles
  • Small refrigeration circulators are optimised for laboratories, they